siRNA Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013120.2 | NP_037252.1 | 16, 17 | 1594 | ||
| GenBank mRNA | BC081843.1 | - | 16, 17 | 1594 | 7096237 | |
| KJ026952.1 | - | 16, 17 | 1417 | |||
| X68497.1 | - | 16, 17 | 1438 | |||

Gckr
Genomic Map