siRNA Details
| Gene Symbol: Fn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019143.2 | NP_062016.2 | 25 | 4095 | ||
| XM_006245150.2 | XP_006245212.1 | 25 | 4097 | |||
| XM_006245151.2 | XP_006245213.1 | 25 | 4096 | |||
| XM_006245154.2 | XP_006245216.1 | 25 | 4097 | |||
| XM_006245155.2 | XP_006245217.1 | 22 | 3469 | |||
| GenBank mRNA | X15906.1 | - | 25 | 4093 | ||

Fn1
Genomic Map