siRNA Details
| Gene Symbol: Cd38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013127.1 | NP_037259.1 | 6 | 698 | ||
| GenBank mRNA | D29646.1 | - | 6 | 698 | ||
| D30795.1 | - | 6 | 693 | |||
| FQ212353.1 | - | 6 | 703 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd38| Gene Symbol: Cd38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013127.1 | NP_037259.1 | 6 | 698 | ||
| GenBank mRNA | D29646.1 | - | 6 | 698 | ||
| D30795.1 | - | 6 | 693 | |||
| FQ212353.1 | - | 6 | 703 | |||