siRNA Details
| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013129.2 | NP_037261.1 | 4 | 277 | ||
| GenBank mRNA | AF015719.1 | - | 4 | 277 | ||
| BC097340.1 | - | 5 | 1081 | |||
| U69272.1 | - | 822 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il15| Gene Symbol: Il15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013129.2 | NP_037261.1 | 4 | 277 | ||
| GenBank mRNA | AF015719.1 | - | 4 | 277 | ||
| BC097340.1 | - | 5 | 1081 | |||
| U69272.1 | - | 822 | ||||