siRNA Details
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013131.1 | NP_037263.1 | 2 | 838 | ||
GenBank mRNA | M36074.1 | - | 2 | 838 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nr3c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013131.1 | NP_037263.1 | 2 | 838 | ||
GenBank mRNA | M36074.1 | - | 2 | 838 |