siRNA Details
| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013132.1 | NP_037264.1 | 12 | 898 | ||
| XM_006232268.2 | XP_006232330.1 | 12 | 1033 | |||
| GenBank mRNA | AF051895.1 | - | 12 | 953 | ||
| BC081855.1 | - | 12 | 951 | 7192977 | ||
| FQ213209.1 | - | 13 | 1028 | |||
| FQ214176.1 | - | 12 | 930 | |||
| FQ229127.1 | - | 12 | 886 | |||
| M21730.1 | - | 12 | 898 | |||

Anxa5
Genomic Map