siRNA Details
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013132.1 | NP_037264.1 | 10 | 722 | ||
XM_006232268.2 | XP_006232330.1 | 10 | 857 | |||
GenBank mRNA | AF051895.1 | - | 10 | 777 | ||
BC081855.1 | - | 10 | 775 | 7192977 | ||
FQ213209.1 | - | 11 | 852 | |||
FQ214176.1 | - | 10 | 754 | |||
FQ229127.1 | - | 10 | 710 | |||
M21730.1 | - | 10 | 722 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
