siRNA Details
| Gene Symbol: Rasa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013135.1 | NP_037267.1 | 16 | 2054 | ||
| GenBank mRNA | FQ227936.1 | - | 3 | 285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasa1| Gene Symbol: Rasa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013135.1 | NP_037267.1 | 16 | 2054 | ||
| GenBank mRNA | FQ227936.1 | - | 3 | 285 | ||