siRNA Details
Gene Symbol: Mak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013136.1 | NP_037268.1 | 5 | 677 | ||
GenBank mRNA | BC078887.1 | - | 5 | 665 | 7111717 | |
M35862.1 | - | 5 | 677 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013136.1 | NP_037268.1 | 5 | 677 | ||
GenBank mRNA | BC078887.1 | - | 5 | 665 | 7111717 | |
M35862.1 | - | 5 | 677 |