siRNA Details
| Gene Symbol: Col10a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001053056.6 | XP_001053056.5 | 3 | 5245 | ||
| XM_008773017.1 | XP_008771239.1 | 2 | 5762 | |||
| XM_008773018.1 | XP_008771240.1 | 3 | 5718 | |||
| GenBank mRNA | AJ131848.1 | - | 1 | 637 | ||

Col10a1
Genomic Map