siRNA Details
| Gene Symbol: Ppard | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013141.2 | NP_037273.2 | 6 | 790 | ||
| XM_006256166.2 | XP_006256228.1 | 6 | 1602 | |||
| XM_006256167.2 | XP_006256229.1 | 5 | 1709 | |||
| XM_006256168.2 | XP_006256230.1 | 5 | 2604 | |||
| XM_008772797.1 | XP_008771019.1 | 5 | 1837 | |||
| GenBank mRNA | AJ306400.1 | - | 4 | 556 | ||
| U40064.1 | - | 6 | 811 | |||
| U75918.1 | - | 6 | 804 | |||

Ppard
Genomic Map