siRNA Details
| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013143.1 | NP_037275.1 | 7 | 582 | ||
| XM_008766837.1 | XP_008765059.1 | 7 | 621 | |||
| XM_008766838.1 | XP_008765060.1 | 5 | 443 | |||
| GenBank mRNA | BC081834.1 | - | 7 | 597 | 7128404 | |
| S43408.1 | - | 7 | 582 | |||

Mep1a
Genomic Map