siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 15 | 1732 | ||
| GenBank mRNA | X62894.1 | - | 15 | 1732 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clcn1| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 15 | 1732 | ||
| GenBank mRNA | X62894.1 | - | 15 | 1732 | ||