siRNA Details
| Gene Symbol: Clcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013147.1 | NP_037279.1 | 11 | 1264 | ||
| XM_008762873.1 | XP_008761095.1 | 10 | 2078 | |||
| GenBank mRNA | AY112736.1 | - | 10 | 1013 | ||
| AY112737.1 | - | 12 | 1174 | |||
| AY112738.1 | - | 11 | 1095 | |||
| AY112739.1 | - | 10 | 966 | |||
| AY112740.1 | - | 11 | 1092 | |||
| X62894.1 | - | 11 | 1264 | |||

Clcn1
Genomic Map