siRNA Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013156.2 | NP_037288.1 | 7 | 931 | ||
| GenBank mRNA | AB630323.1 | - | 6 | 872 | ||
| BC063175.1 | - | 7 | 946 | 6918389 | ||
| FQ209613.1 | - | 7 | 943 | |||
| FQ218649.1 | - | 7 | 940 | |||
| FQ228725.1 | - | 7 | 942 | |||
| FQ228844.1 | - | 7 | 942 | |||
| Y00697.1 | - | 7 | 935 | |||

Ctsl
Genomic Map