siRNA Details
| Gene Symbol: Ass1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013157.3 | NP_037289.1 | 12 | 1119 | ||
| XM_006233911.2 | XP_006233973.1 | 14 | 1270 | |||
| GenBank mRNA | BC063146.1 | - | 14 | 1260 | 6888536 | |
| M36708.1 | - | 12 | 1119 | |||
| X12459.1 | - | 12 | 1119 | |||

Ass1
Genomic Map