siRNA Details
| Gene Symbol: Ide | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013159.1 | NP_037291.1 | 11 | 1402 | ||
| XM_006231313.2 | XP_006231375.1 | 11 | 2173 | |||
| XM_006231314.2 | XP_006231376.1 | 11 | 1386 | |||
| XM_006231315.2 | XP_006231377.1 | 11 | 1456 | |||
| XM_008760362.1 | XP_008758584.1 | 11 | 2143 | |||
| GenBank mRNA | X67269.1 | - | 11 | 1402 | ||

Ide
Genomic Map