siRNA Details
| Gene Symbol: Pnlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013161.2 | NP_037293.1 | 8 | 815 | ||
| GenBank mRNA | D88534.1 | - | 7 | 787 | ||
| D88535.1 | - | 7 | 787 | |||
| M58369.1 | - | 8 | 816 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnlip| Gene Symbol: Pnlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013161.2 | NP_037293.1 | 8 | 815 | ||
| GenBank mRNA | D88534.1 | - | 7 | 787 | ||
| D88535.1 | - | 7 | 787 | |||
| M58369.1 | - | 8 | 816 | |||