siRNA Details
| Gene Symbol: Zeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013164.1 | NP_037296.1 | 6 | 942 | ||
| GenBank mRNA | AY325179.1 | - | 6 | 942 | ||
| U51583.1 | - | 1189 | ||||
| U51584.1 | - | 2330 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zeb1| Gene Symbol: Zeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013164.1 | NP_037296.1 | 6 | 942 | ||
| GenBank mRNA | AY325179.1 | - | 6 | 942 | ||
| U51583.1 | - | 1189 | ||||
| U51584.1 | - | 2330 | ||||