siRNA Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013167.2 | NP_037299.1 | 7 | 987 | ||
| GenBank mRNA | AB006614.1 | - | 6 | 859 | ||
| AF030163.1 | - | 6 | 874 | |||
| AF035943.1 | - | 6 | 870 | |||
| BC072546.1 | - | 7 | 987 | 7105743 | ||
| U92069.1 | - | 7 | 994 | |||

Ucp3
Genomic Map