siRNA Details
Gene Symbol: Hmbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013168.2 | NP_037300.2 | 4 | 321 | ||
XM_006242896.1 | XP_006242958.1 | 4 | 407 | |||
XM_006242897.1 | XP_006242959.1 | 4 | 407 | |||
XM_008766148.1 | XP_008764370.1 | 4 | 407 | |||
XM_008766149.1 | XP_008764371.1 | 4 | 344 | |||
GenBank mRNA | BC088162.1 | - | 4 | 321 | 7375228 | |
FQ226062.1 | - | 4 | 244 | |||
FQ231091.1 | - | 4 | 305 | |||
FQ234870.1 | - | 3 | 393 | |||
X06827.1 | - | 4 | 196 | |||
Y12006.1 | - | 4 | 191 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013551, NM_001110251, AK167702, AK166734, AK143558, and others.
Panther Classification:
Gene Ontology Categories:
