siRNA Details
Gene Symbol: Ifng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138880.2 | NP_620235.1 | 3 | 287 | ||
GenBank mRNA | AF010466.1 | - | 3 | 287 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138880.2 | NP_620235.1 | 3 | 287 | ||
GenBank mRNA | AF010466.1 | - | 3 | 287 |