siRNA Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033680.2 | NP_001028852.2 | 5 | 679 | ||
| XM_008765352.1 | XP_008763574.1 | 2 | 661 | |||
| XM_008765353.1 | XP_008763575.1 | 2 | 664 | |||
| GenBank mRNA | AJ617615.1 | - | 5 | 679 | ||
| CB731032.1 | - | 1 | 61 | |||
| DQ181550.2 | - | 2 | 347 | |||
| X52772.1 | - | 5 | 862 | |||

Syt1
Genomic Map