siRNA Details
| Gene Symbol: Scg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013175.3 | NP_037307.1 | 4 | 473 | ||
| XM_006234701.2 | XP_006234763.1 | 4 | 546 | |||
| GenBank mRNA | BC061560.1 | - | 4 | 477 | 6918342 | |
| FQ212495.1 | - | 4 | 473 | |||
| FQ213081.1 | - | 4 | 499 | |||
| FQ213100.1 | - | 4 | 499 | |||
| M63901.1 | - | 4 | 471 | |||

Scg5
Genomic Map