siRNA Details
| Gene Symbol: Tcf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013176.2 | NP_037308.1 | 7, 8 | 726 | ||
| XM_006243345.1 | XP_006243407.1 | 7, 8 | 584 | |||
| XM_006243346.1 | XP_006243408.1 | 7, 8 | 584 | |||
| XM_006243347.1 | XP_006243409.1 | 7, 8 | 584 | |||
| XM_006243348.1 | XP_006243410.1 | 7, 8 | 584 | |||
| XM_006243349.1 | XP_006243411.1 | 7, 8 | 584 | |||
| XM_006243350.1 | XP_006243412.1 | 7, 8 | 584 | |||
| XM_006243351.1 | XP_006243413.1 | 5, 6 | 437 | |||
| XM_008766218.1 | XP_008764440.1 | 5, 6 | 4265 | |||
| GenBank mRNA | L09656.1 | - | 7, 8 | 726 | ||

Tcf12
Genomic Map