siRNA Details
| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013177.2 | NP_037309.1 | 3 | 432 | ||
| GenBank mRNA | BC061792.1 | - | 3 | 380 | 5597941 | |
| FM056810.1 | - | 3 | 432 | |||
| M18467.1 | - | 3 | 403 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Got2| Gene Symbol: Got2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013177.2 | NP_037309.1 | 3 | 432 | ||
| GenBank mRNA | BC061792.1 | - | 3 | 380 | 5597941 | |
| FM056810.1 | - | 3 | 432 | |||
| M18467.1 | - | 3 | 403 | |||