siRNA Details
| Gene Symbol: Mep1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013183.2 | NP_037315.1 | 13 | 1841 | ||
| XM_006254458.2 | XP_006254520.1 | 12 | 1982 | |||
| XM_006254459.2 | XP_006254521.1 | 13 | 1810 | |||
| GenBank mRNA | BC081833.1 | - | 13 | 1817 | 7108543 | |
| M88601.1 | - | 13 | 1842 | |||

Mep1b
Genomic Map