siRNA Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100821.1 | NP_001094291.1 | 4, 5 | 357 | ||
| XM_006228904.2 | XP_006228966.1 | 4, 5 | 509 | |||
| GenBank mRNA | BC162008.1 | - | 4, 5 | 384 | 9090267 | |
| D14015.1 | - | 4, 5 | 374 | |||
| EV771022.1 | - | 4, 5 | 391 | |||

Ccne1
Genomic Map