siRNA Details
| Gene Symbol: Ccne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100821.1 | NP_001094291.1 | 9 | 927 | ||
| XM_006228904.2 | XP_006228966.1 | 9 | 1079 | |||
| GenBank mRNA | BC162008.1 | - | 9 | 954 | 9090267 | |
| D14015.1 | - | 9 | 944 | |||
| DY561075.1 | - | 4 | 363 | |||

Ccne1
Genomic Map