siRNA Details
| Gene Symbol: Hck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013185.3 | NP_037317.2 | 2, 3 | 376 | ||
| GenBank mRNA | BC078890.1 | - | 2, 3 | 376 | 7121407 | |
| M83666.1 | - | 2, 3 | 298 | |||
| S74141.1 | - | 2, 3 | 298 | |||
| X62345.1 | - | 2, 3 | 327 | |||

Hck
Genomic Map