siRNA Details
| Gene Symbol: Kcnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013186.1 | NP_037318.1 | 2 | 1429 | ||
| GenBank mRNA | X16476.1 | - | 2 | 1429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnb1| Gene Symbol: Kcnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013186.1 | NP_037318.1 | 2 | 1429 | ||
| GenBank mRNA | X16476.1 | - | 2 | 1429 | ||