siRNA Details
| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013195.1 | NP_037327.1 | 4 | 329 | ||
| XM_006241970.2 | XP_006242032.1 | 4 | 758 | |||
| GenBank mRNA | M55050.1 | - | 4 | 329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il2rb| Gene Symbol: Il2rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013195.1 | NP_037327.1 | 4 | 329 | ||
| XM_006241970.2 | XP_006242032.1 | 4 | 758 | |||
| GenBank mRNA | M55050.1 | - | 4 | 329 | ||