siRNA Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013196.1 | NP_037328.1 | 8 | 1630 | ||
| XM_006242151.2 | XP_006242213.1 | 7 | 2586 | |||
| XM_006242152.2 | XP_006242214.1 | 9 | 1858 | |||
| XM_006242153.2 | XP_006242215.1 | 9 | 1573 | |||
| XM_006242154.2 | XP_006242216.1 | 9 | 1627 | |||
| XM_006242156.2 | XP_006242218.1 | 8 | 1457 | |||
| XM_008765676.1 | XP_008763898.1 | 9 | 1650 | |||
| GenBank mRNA | M88592.1 | - | 8 | 1630 | ||

Ppara
Genomic Map