siRNA Details
| Gene Symbol: Maob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013198.1 | NP_037330.1 | 3 | 260 | ||
| GenBank mRNA | BC078886.1 | - | 3 | 267 | ||
| BC089814.1 | - | 3 | 225 | 7323595 | ||
| FQ210326.1 | - | 3 | 321 | |||
| M23601.1 | - | 3 | 260 | |||

Maob
Genomic Map