siRNA Details
| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019145.1 | NP_062018.1 | 12 | 1562 | ||
| GenBank mRNA | X74834.1 | - | 12 | 1562 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrng| Gene Symbol: Chrng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019145.1 | NP_062018.1 | 12 | 1562 | ||
| GenBank mRNA | X74834.1 | - | 12 | 1562 | ||