siRNA Details
| Gene Symbol: Acot7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146061.1 | NP_001139533.1 | 3 | 446 | ||
| NM_013214.4 | NP_037346.2 | 3 | 374 | |||
| XM_006239447.2 | XP_006239509.1 | 3 | 474 | |||
| XM_006239448.1 | XP_006239510.1 | 3 | 382 | |||
| GenBank mRNA | BC087716.1 | - | 3 | 326 | 7316866 | |
| D88891.1 | - | 3 | 446 | |||
| Y09332.1 | - | 3 | 274 | |||

Acot7
Genomic Map