siRNA Details
| Gene Symbol: Cartpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017110.1 | NP_058806.1 | 2 | 255 | ||
| XM_006231843.2 | XP_006231905.1 | 2 | 640 | |||
| GenBank mRNA | U10071.1 | - | 2 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cartpt| Gene Symbol: Cartpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017110.1 | NP_058806.1 | 2 | 255 | ||
| XM_006231843.2 | XP_006231905.1 | 2 | 640 | |||
| GenBank mRNA | U10071.1 | - | 2 | 255 | ||