siRNA Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033237.1 | NP_150240.1 | 5 | 435 | ||
| GenBank mRNA | J03624.1 | - | 5 | 435 | ||
| M18102.1 | - | 5 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033237.1 | NP_150240.1 | 5 | 435 | ||
| GenBank mRNA | J03624.1 | - | 5 | 435 | ||
| M18102.1 | - | 5 | 415 | |||