siRNA Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172008.2 | NP_742005.1 | 11 | 1431 | ||
| XM_008767679.1 | XP_008765901.1 | 11 | 1552 | |||
| XM_008767680.1 | XP_008765902.1 | 11 | 1471 | |||
| GenBank mRNA | FQ210638.1 | - | 11 | 1457 | ||
| L18889.1 | - | 10 | 1328 | |||

Canx
Genomic Map