siRNA Details
| Gene Symbol: Ctxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109935.1 | NP_001103405.1 | 2 | 839 | ||
| GenBank mRNA | L15011.1 | - | 2 | 837 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctxn1| Gene Symbol: Ctxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109935.1 | NP_001103405.1 | 2 | 839 | ||
| GenBank mRNA | L15011.1 | - | 2 | 837 | ||