siRNA Details
| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240666.2 | XP_006240728.1 | 6 | 1037 | ||
| XM_006240667.2 | XP_006240729.1 | 6 | 1036 | |||
| GenBank mRNA | U70050.1 | - | 6 | 849 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jag2| Gene Symbol: Jag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240666.2 | XP_006240728.1 | 6 | 1037 | ||
| XM_006240667.2 | XP_006240729.1 | 6 | 1036 | |||
| GenBank mRNA | U70050.1 | - | 6 | 849 | ||