siRNA Details
| Gene Symbol: Ucn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019150.1 | NP_062023.1 | 2 | 536 | ||
| GenBank mRNA | U33935.1 | - | 2 | 536 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ucn| Gene Symbol: Ucn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019150.1 | NP_062023.1 | 2 | 536 | ||
| GenBank mRNA | U33935.1 | - | 2 | 536 | ||