siRNA Details
| Gene Symbol: Capn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019152.2 | NP_062025.1 | 7 | 831 | ||
| XM_006230706.2 | XP_006230768.1 | 7 | 890 | |||
| XM_008760088.1 | XP_008758310.1 | 7 | 1462 | |||
| XM_008760089.1 | XP_008758311.1 | 7 | 1142 | |||
| GenBank mRNA | BC061880.1 | - | 7 | 831 | 5599301 | |
| U53858.1 | - | 7 | 821 | |||

Capn1
Genomic Map