siRNA Details
| Gene Symbol: Capns1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017118.1 | NP_058814.1 | 9 | 825 | ||
| GenBank mRNA | BC098068.1 | - | 9 | 825 | 7462537 | |
| FQ229527.1 | - | 508 | ||||
| U10861.1 | - | 514 | ||||
| U53859.1 | - | 8 | 761 | |||
| Gene Symbol: LOC100912380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228796.2 | XP_006228858.2 | 601 | |||

Capns1
Genomic Map