siRNA Details
| Gene Symbol: Ccng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105725.2 | NP_001099195.1 | 6 | 868 | ||
| GenBank mRNA | BC169085.1 | - | 6 | 868 | 7938657 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccng2| Gene Symbol: Ccng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105725.2 | NP_001099195.1 | 6 | 868 | ||
| GenBank mRNA | BC169085.1 | - | 6 | 868 | 7938657 | |