siRNA Details
| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271073.1 | NP_001258002.1 | 5 | 328 | ||
| NM_001271074.1 | NP_001258003.1 | 5 | 400 | |||
| NM_001271077.1 | NP_001258006.1 | 5 | 400 | |||
| NM_019154.1 | NP_062027.1 | 5 | 391 | |||
| GenBank mRNA | U01245.2 | - | 4 | 359 | ||
| U51195.1 | - | 5 | 401 | |||
| U60562.1 | - | 5 | 329 | |||
| U67130.1 | - | 5 | 391 | |||

Amelx
Genomic Map