siRNA Details
| Gene Symbol: Hpca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017122.1 | NP_058818.1 | 1 | 313 | ||
| XM_006238920.1 | XP_006238982.1 | 2 | 258 | |||
| XM_006238921.2 | XP_006238983.1 | 2 | 215 | |||
| GenBank mRNA | AY442172.1 | - | 1 | 140 | ||
| BC087632.1 | - | 1 | 168 | 7319524 | ||
| D12573.1 | - | 1 | 313 | |||

Hpca
Genomic Map