siRNA Details
| Gene Symbol: Syn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034020.1 | NP_001029192.1 | 10, 11 | 1449 | ||
| NM_019159.1 | NP_062032.1 | 10, 11 | 1449 | |||
| GenBank mRNA | M27925.1 | - | 10, 11 | 1449 | ||
| M27926.1 | - | 10, 11 | 1449 | |||

Syn2
Genomic Map