siRNA Details
| Gene Symbol: Cd69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134327.1 | NP_599154.1 | 2 | 227 | ||
| GenBank mRNA | BC088219.1 | - | 2 | 227 | 7384461 | |
| GU357488.1 | - | 2 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd69| Gene Symbol: Cd69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134327.1 | NP_599154.1 | 2 | 227 | ||
| GenBank mRNA | BC088219.1 | - | 2 | 227 | 7384461 | |
| GU357488.1 | - | 2 | 163 | |||