siRNA Details
Gene Symbol: Cd69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134327.1 | NP_599154.1 | 2, 3 | 251 | ||
GenBank mRNA | BC088219.1 | - | 2, 3 | 251 | 7384461 | |
GU357488.1 | - | 2, 3 | 187 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134327.1 | NP_599154.1 | 2, 3 | 251 | ||
GenBank mRNA | BC088219.1 | - | 2, 3 | 251 | 7384461 | |
GU357488.1 | - | 2, 3 | 187 |